Learn More
3,5-Bis(1,1-dimethylethyl)-4-hydroxy-3-hydroxy-2,2-bis(hydroxymethyl)propyl Benzenepropanoic Acid Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Bis(1,1-dimethylethyl)-4-hydroxy-3-hydroxy-2,2-bis(hydroxymethyl)propyl Benzenepropanoic Acid Ester |
| CAS | 26347-98-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H36 O6 |
| Quantity | 250 mg |
| Synonym | Agidol 110 |
| InChI Formula | InChI=1S/C22H36O6/c1-20(2,3)16-9-15(10-17(19(16)27)21(4,5)6)7-8-18(26)28-14-22(11-23,12-24)13-25/h9-10,23-25,27H,7-8,11-14H2,1-6H3 |
| SMILES | CC(C)(C)c1cc(CCC(=O)OCC(CO)(CO)CO)cc(c1O)C(C)(C)C |
| IUPAC Name | [3-hydroxy-2,2-bis(hydroxymethyl)propyl] 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate |
| Molecular Weight (g/mol) | 396.5176 |
| Show More |