Learn More
3',5'-Bis-O-[(1,1-dimethylethyl)dimethylsilyl]thymidine-alpha-13C, TRC
Specifications
Specifications
| Chemical Name or Material | 3',5'-Bis-O-[(1,1-dimethylethyl)dimethylsilyl]thymidine-Alpha-13C |
| CAS | 210101-69-2 |
| Molecular Formula | 13C C21 H42 N2 O5 Si2 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C22H42N2O5Si2/c1-15-13-24(20(26)23-19(15)25)18-12-16(29-31(10,11)22(5,6)7)17(28-18)14-27-30(8,9)21(2,3)4/h13,16-18H,12,14H2,1-11H3,(H,23,25,26)/t16-,17+,18+/m0/s1/i1+1 |
| SMILES | [13CH3]C1=CN([C@H]2C[C@H](O[Si](C)(C)C(C)(C)C)[C@@H](CO[Si](C)(C)C(C)(C)C)O2)C(=O)NC1=O |
| IUPAC Name | 1-[(2R,4S,5R)-4-[tert-butyl(dimethyl)silyl]oxy-5-[[tert-butyl(dimethyl)silyl]oxymethyl]oxolan-2-yl]-5-(113C)methylpyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 471.743 |
| Formula Weight | 471.267 |