Learn More
3,5-Di(1-pyrenyl)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Di(1-pyrenyl)pyridine |
| CAS | 1246467-58-2 |
| Molecular Formula | C37H21N |
| Quantity | 10 mg |
| Synonym | 3,5-Di-1-pyrenylpyridine |
| InChI Formula | InChI=1S/C37H21N/c1-3-22-7-9-26-11-15-30(32-17-13-24(5-1)34(22)36(26)32)28-19-29(21-38-20-28)31-16-12-27-10-8-23-4-2-6-25-14-18-33(31)37(27)35(23)25/h1-21H |
| SMILES | c1cc2ccc3ccc(c4cncc(c4)c5ccc6ccc7cccc8ccc5c6c78)c9ccc(c1)c2c39 |
| Molecular Weight (g/mol) | 479.57 |
| Formula Weight | 479.1674 |