missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzoic acid, 99%, Thermo Scientific™

Catalog Number 15545076
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
618-58-6
C7H4Br2O2
279.92
MFCD00051758
SFTFNJZWZHASAQ-UHFFFAOYSA-N
benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h
12063
3,5-dibromobenzoic acid
OC(=O)C1=CC(Br)=CC(Br)=C1
This item is not returnable. View return policy
618-58-6
C7H4Br2O2
279.92
MFCD00051758
SFTFNJZWZHASAQ-UHFFFAOYSA-N
benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h
12063
3,5-dibromobenzoic acid
OC(=O)C1=CC(Br)=CC(Br)=C1
Melting Point 212.0°C to 214.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula Br2C6H3CO2H
Quantity 1 g
Formula Weight 279.91
Percent Purity 99%
Chemical Name or Material 3,5-Dibromobenzoic acid
compliance-icons
Product Identifier
  • 3,5-Dibromobenzoic acid, 99%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'