Learn More
3,5-Dichloro-2,6-dimethylpyridin-4-ol, 95%, BLD Pharm™
Product Code. 30961308
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
100 g
Unit Size:
100g
100mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30961308 100 mg 100mg
30950034 5 g 5g
30961158 100 g 100g
30950642 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30961308 Supplier BLD Pharm Supplier No. BD121981100mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 2971-90-6
Molecular Formula C7H7Cl2NO
MDL Number MFCD22123390
Quantity 100 mg
SMILES OC1=C(Cl)C(C)=NC(C)=C1Cl
Molecular Weight (g/mol) 192.04
Percent Purity 95%
Chemical Name or Material 3,5-Dichloro-2,6-dimethylpyridin-4-ol