missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-N-methylbenzylamine, 95%, Thermo Scientific™

Catalog Number p-4228590
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
90390-21-9
C8H9Cl2N
190.07
SSTJQZMMDIIKBR-UHFFFAOYSA-N
n-methyl-3,5-dichlorobenzylamine, 3,5-dichlorophenyl methyl methyl amine, 3,5-dichloro-n-methylbenzylamine, 1-3,5-dichlorophenyl-n-methylmethanamine, benzenemethanamine, 3,5-dichloro-n-methyl, 1-3,5-dichlorophenyl-n-methyl-methanamine, benzylamine, n-methyl-3,5-dichloro, 90389-22-3 hydrochloride, 3,5-dichloro-n-methyl-benzenemethanamine
146163
1-(3,5-dichlorophenyl)-N-methylmethanamine
CNCC1=CC(=CC(=C1)Cl)Cl
This item is not returnable. View return policy
90390-21-9
C8H9Cl2N
190.07
SSTJQZMMDIIKBR-UHFFFAOYSA-N
n-methyl-3,5-dichlorobenzylamine, 3,5-dichlorophenyl methyl methyl amine, 3,5-dichloro-n-methylbenzylamine, 1-3,5-dichlorophenyl-n-methylmethanamine, benzenemethanamine, 3,5-dichloro-n-methyl, 1-3,5-dichlorophenyl-n-methyl-methanamine, benzylamine, n-methyl-3,5-dichloro, 90389-22-3 hydrochloride, 3,5-dichloro-n-methyl-benzenemethanamine
146163
1-(3,5-dichlorophenyl)-N-methylmethanamine
CNCC1=CC(=CC(=C1)Cl)Cl
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 190.07
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3, 5-Dichloro-N-methylbenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Dichloro-N-methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'