Learn More
3,5-Difluoro-D-phenylalanine, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Difluoro-D-phenylalanine |
| CAS | 266360-63-8 |
| Molecular Formula | C21 H24 O12 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C21H24O12/c1-10(22)28-9-16-17(29-11(2)23)18(30-12(3)24)19(31-13(4)25)21(33-16)32-15-8-6-5-7-14(15)20(26)27/h5-8,16-19,21H,9H2,1-4H3,(H,26,27)/t16?,17-,18?,19?,21+/m0/s1 |
| SMILES | CC(=O)OCC1O[C@@H](Oc2ccccc2C(=O)O)C(OC(=O)C)C(OC(=O)C)[C@H]1OC(=O)C |
| IUPAC Name | 2-[(2S,5S)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxybenzoic acid |
| Molecular Weight (g/mol) | 468.408 |
| Formula Weight | 468.127 |