missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-4-pyridone-1-acetic acid, 99%, Thermo Scientific™

Product Code. 11471667
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11471667

Brand: Thermo Scientific Alfa Aesar B25646.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-29-1
Molecular Formula C7H5I2NO3
Molecular Weight (g/mol) 404.93
MDL Number MFCD00006185
InChI Key PVBALTLWZVEAIO-UHFFFAOYSA-N
Synonym pelvirinic acid, 3,5-diiodo-4-pyridone-1-acetic acid, umbradilic acid, diodone acid, unii-nlx9tz649p, 1 4h-pyridineacetic acid, 3,5-diiodo-4-oxo, nlx9tz649p, 3,5-diiodo-4-oxopyridin-1 4h-yl acetic acid, 2-3,5-diiodo-4-oxopyridin-1 4h-yl acetic acid, 3,5-diiodo-4-pyridone-n-acetic acid
PubChem CID 9304
IUPAC Name 2-(3,5-diiodo-4-oxopyridin-1-yl)acetic acid
SMILES C1=C(C(=O)C(=CN1CC(=O)O)I)I

Specifications

Melting Point 244°C (decomposition)
Quantity 1 g
Beilstein 163586
Sensitivity Light sensitive
Formula Weight 404.93
Percent Purity 99%
Chemical Name or Material 3,5-Diiodo-4-pyridone-1-acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.