Learn More
3,5-Dimethoxybenzyl Bromide, TRC
Product Code. 30376703
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376703 5 g
30386858 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30376703 Supplier Toronto Research Chemicals Supplier No. TRCD4608355G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 877-88-3
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.09
Synonym 1-(Bromomethyl)-3,5-dimethoxy-benzene, 5-(Bromomethyl)-1,3-dimethoxybenzene, alpha-Bromo-3,5-dimethoxytoluene
SMILES COc1cc(CBr)cc(OC)c1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C9H11BrO2/c1-11-8-3-7(6-10)4-9(5-8)12-2/h3-5H,6H2,1-2H3
Formula Weight 229.9942
Chemical Name or Material 3,5-Dimethoxybenzyl Bromide