Learn More
3,5-Dimethyl-1-phenyl-1H-pyrazole, 97%, Thermo Scientific™
Product Code. 11467166
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467166 5 g 5g
11477166 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11467166 Supplier Thermo Scientific Alfa Aesar Supplier No. B25489.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1131-16-4
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD00020728
InChI Key ULPMPUPEFBDQQA-UHFFFAOYSA-N
PubChem CID 70800
IUPAC Name 3,5-dimethyl-1-phenylpyrazole
SMILES CC1=CC(=NN1C2=CC=CC=C2)C
Specifications
Boiling Point 144°C to 145°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 132902
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole, 3,5-dimethyl-1-phenyl, dimethylphenylpyrazole, unii-0sb52bxx7m, 1h-pyrazole,3,5-dimethyl-1-phenyl, 0sb52bxx7m, 1,3-dimethyl-5-phenylpyrazole, phenyldimethylpyrazole, enamine_001158, pyrazole, dimethylphenyl
Formula Weight 172.23
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-1-phenyl-1H-pyrazole