missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyladamantane-1-methanol, 97%, Thermo Scientific™

Catalog Number p-7052611
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
26919-42-6
C13H22O
194.318
MFCD00188062
RVWLWJAOIBEWAV-UHFFFAOYSA-N
3,5-dimethyladamantan-1-yl methanol, 3,5-dimethyl-1-adamantanemethanol, tricyclo 3.3.1.13,7 decane-1-methanol, 3,5-dimethyl, 3,5-dimethyladamantanyl methan-1-ol, acmc-1cfe2, 1r,3r,5s,7r-3,5-dimethyladamantan-1-yl methanol, 3,5-dimethyl-1-adamantyl methanol, 3,5-dimethyladamant-1-yl methanol, 3,5-dimethyl-1-hydroxymethyladamantane, 1-hydroxymethyl-3,5-dimethyladamantane
4050097
(3,5-dimethyl-1-adamantyl)methanol
CC12CC3CC(C1)(CC(C3)(C2)CO)C
This item is not returnable. View return policy
26919-42-6
C13H22O
194.318
MFCD00188062
RVWLWJAOIBEWAV-UHFFFAOYSA-N
3,5-dimethyladamantan-1-yl methanol, 3,5-dimethyl-1-adamantanemethanol, tricyclo 3.3.1.13,7 decane-1-methanol, 3,5-dimethyl, 3,5-dimethyladamantanyl methan-1-ol, acmc-1cfe2, 1r,3r,5s,7r-3,5-dimethyladamantan-1-yl methanol, 3,5-dimethyl-1-adamantyl methanol, 3,5-dimethyladamant-1-yl methanol, 3,5-dimethyl-1-hydroxymethyladamantane, 1-hydroxymethyl-3,5-dimethyladamantane
4050097
(3,5-dimethyl-1-adamantyl)methanol
CC12CC3CC(C1)(CC(C3)(C2)CO)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 55°C to 56°C
Color White
Boiling Point 114°C (2.25 mmHg)
Quantity 1 g
Formula Weight 194.32
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,5-Dimethyladamantane-1-methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'