Learn More
3,5-Xylenol Phosphate, TRC
Product Code. 30449310
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449310 10 g 10g
30451791 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30449310 Supplier Toronto Research Chemicals Supplier No. TRCX74955010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25653-16-1
Molecular Formula C24H27O4P
Molecular Weight (g/mol) 410.44
Synonym Tri-3,5-xylyl Phosphate, Phenol, 3,5-dimethyl-, 1,1',1' '-phosphate, 3,5-Xylenol, phosphate (3:1) (8CI), 3,5-Xylyl phosphate ((C8H9O)3PO) (6CI), 3,5-Xylyl phosphate (7CI), Phenol, 3,5-dimethyl-, phosphate (3:1) (9CI), NSC 66515, Tri-3,5-xylenyl phosphate, Tri-3,5-xylyl phosphate, Tris(3,5-dimethylphenyl) phosphate, Tri-3,5-xylyl phosphate, Tris(3,5-dimethylphenyl) Phosphate, 3,5-Dimethylphenol 1,1',1' '-Phosphate, 3,5-Dimethylphenol Phosphate (3:1)
IUPAC Name tris(3,5-dimethylphenyl) phosphate
SMILES Cc1cc(C)cc(OP(=O)(Oc2cc(C)cc(C)c2)Oc3cc(C)cc(C)c3)c1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C24H27O4P/c1-16-7-17(2)11-22(10-16)26-29(25,27-23-12-18(3)8-19(4)13-23)28-24-14-20(5)9-21(6)15-24/h7-15H,1-6H3
Formula Weight 410.1647
Percent Purity >95%
Chemical Name or Material 3,5-Xylenol Phosphate
Grade HPLC