Learn More
3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 520-45-6 |
| Molecular Formula | C8H8O4 |
| MDL Number | MFCD00066709 |
| Quantity | 100 g |
| SMILES | CC(C1C(C=C(C)OC1=O)=O)=O |
| Molecular Weight (g/mol) | 168.15 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione |