missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-1,4-dimethyl-5H-pyrido[4,3-b]indole-13C2,15N Acetate, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | 3-Amino-1,4-dimethyl-5h-pyrido[4,3-b]indole-13C2,15N Acetate |
| CAS | 1217002-93-1 |
| Molecular Formula | C1313C2H17N215NO2 |
| Quantity | 0.5 mg |
| Synonym | 3-(Amino-15N)-1-methyl-4-(methyl-13C)-5H-pyrido[4,3-b]indole-3-13C monoacetate, 3-Amino-1,4-dimethyl-5H-pyrido[4,3-b]indole-13C2,15N acetate (3-13C,4-methyl-13C,3-amino-15N), Trp-P 1-13C2,15N HOAc, [13C2,15N]Tryptophan p 1 acetate, 3-Amino-1,4-dimethyl-5H-pyrido[4,3-b]indole-13C2,15N acetate |
| InChI Formula | InChI=1S/C13H13N3.C2H4O2/c1-7-12-11(8(2)15-13(7)14)9-5-3-4-6-10(9)16-12;1-2(3)4/h3-6,16H,1-2H3,(H2,14,15);1H3,(H,3,4)/i1+1,13+1,14+1; |
| SMILES | [13CH3]c1[13c]([15NH2])nc(C)c2c3ccccc3[nH]c12.CC(=O)O |
| IUPAC Name | acetic acid;1-methyl-4-(113C)methyl-5H-(613C)pyridino[4,3-b]indol-3-(15N)amine |
| Molecular Weight (g/mol) | 274.29 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction