Learn More
3-Amino-1-adamantanol, ≥98%, Thermo Scientific Chemicals
Catalog Number p-7055531
Click to view available options
Quantity:
5 g
25 g
100 g
702-82-9
C10H17NO
167.252
MFCD01821204
DWPIPTNBOVJYAD-UHFFFAOYSA-N
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
658645
3-aminoadamantan-1-ol
C1C2CC3(CC1CC(C2)(C3)O)N
702-82-9
C10H17NO
167.252
MFCD01821204
DWPIPTNBOVJYAD-UHFFFAOYSA-N
3-amino-1-adamantanol, 3-amino-1-hydroxyadamantane, 1-amino-3-adamantanol, 3-amino-adamantanol, 1-aminoadamantane-3-ol, 3-hydroxy-1-aminoadamantane, 1-amino-3-hydroxyadamantane, tricyclo 3.3.1.13,7 decan-1-ol, 3-amino, 3-azanyladamantan-1-ol, 3-hydroxyadamantane amine
658645
3-aminoadamantan-1-ol
C1C2CC3(CC1CC(C2)(C3)O)N
It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.
Solubility
Soluble in organic solvents,insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 265°C (literature)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2802127
Solubility Information Soluble in organic solvents,insoluble in water.
Formula Weight 167.25
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 3-Amino-1-adamantanol