Learn More
3-Amino-2-methylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog Number p-4380042
Click to view available options
Quantity:
1 g
5 g
882678-96-8
C13H20BNO2
233.12
MFCD11054038
JMKMGPGFYMANCA-UHFFFAOYSA-N
2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2-methylphenylboronic acid, pinacol ester, 3-amino-2-methylphenylboronic acid pinacol ester, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine, 3-amino-2-methylphenyl boronic acid pinacol ester, 2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylamine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine
46738005
2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
CC1=C(C=CC=C1N)B1OC(C)(C)C(C)(C)O1
882678-96-8
C13H20BNO2
233.12
MFCD11054038
JMKMGPGFYMANCA-UHFFFAOYSA-N
2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2-methylphenylboronic acid, pinacol ester, 3-amino-2-methylphenylboronic acid pinacol ester, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine, 3-amino-2-methylphenyl boronic acid pinacol ester, 2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylamine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine
46738005
2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
CC1=C(C=CC=C1N)B1OC(C)(C)C(C)(C)O1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Boiling Point 120°C to 121°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 233.12
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methylphenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Amino-2-methylphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.