Learn More
3-Amino-2-pyrazinecarboxylic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 3-Amino-2-pyrazinecarboxylic Acid |
| CAS | 5424-01-1 |
| Molecular Formula | C5 H5 N3 O2 |
| Quantity | 10 g |
| Synonym | 2-Pyrazinecarboxylic acid, 3-amino-, Pyrazinecarboxylic acid, 3-amino- (7CI,8CI,9CI), Pyrazinoic acid, 3-amino- (6CI), 3-Amino-2-pyrazinecarboxylic acid, 2-Amino-3-carboxypyrazine, 2-Aminopyrazine-3-carboxylic acid, 3-Amino-2-pyrazinoic acid, 3-Aminopyrazine-2-carboxylic acid, 3-Aminopyrazinecarboxylic acid, NSC 13148, NSC 225114 |
| InChI Formula | InChI=1S/C5H5N3O2/c6-4-3(5(9)10)7-1-2-8-4/h1-2H,(H2,6,8)(H,9,10) |
| SMILES | Nc1nccnc1C(=O)O |
| IUPAC Name | 3-aminopyrazine-2-carboxylic acid |
| Molecular Weight (g/mol) | 139.11 |
| Formula Weight | 139.0382 |