missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific™

Product Code. 10791681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10791681 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10791681 Supplier Thermo Scientific Acros Supplier No. 219300250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2840-28-0
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007671
InChI Key DMGFVJVLVZOSOE-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213
PubChem CID 76092
IUPAC Name 3-amino-4-chlorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 214.0°C to 218.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula H2NC6H3(Cl)CO2H
Quantity 25 g
Beilstein 14, 412
Formula Weight 171.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Amino-4-chlorobenzoic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.