Learn More
3-Amino-4-hydroxybenzoic acid, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 1571-72-8 |
| Molecular Formula | C7H7NO3 |
| MDL Number | MFCD00007697 |
| Quantity | 100 g |
| SMILES | NC1=C(O)C=CC(=C1)C(O)=O |
| Molecular Weight (g/mol) | 153.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Amino-4-hydroxybenzoic acid |