Learn More
3-Amino-6-bromo-4-(2-pyridinyl)-2(1H)-quinolinone(>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 3-Amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one |
| CAS | 77616-97-8 |
| Molecular Formula | C14 H10 Br N3 O |
| Quantity | 25 mg |
| Synonym | 3-Amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one, Bromazepam Imp. D (EP) |
| InChI Formula | InChI=1S/C14H10BrN3O/c15-8-4-5-10-9(7-8)12(13(16)14(19)18-10)11-3-1-2-6-17-11/h1-7H,16H2,(H,18,19) |
| SMILES | NC1=C(c2ccccn2)c3cc(Br)ccc3NC1=O |
| IUPAC Name | 3-amino-6-bromo-4-pyridin-2-yl-1H-quinolin-2-one |
| Molecular Weight (g/mol) | 316.15 |
| Formula Weight | 315.0007 |