Learn More
3-Aminobiphenyl, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Aminobiphenyl |
| CAS | 2243-47-2 |
| Molecular Formula | C12 H11 N |
| Quantity | 10 g |
| Synonym | 3-Biphenylamine (7CI,8CI), (1,1'-Biphenyl-3-yl)amine, 3-Amino-1,1'-biphenyl, 3-Aminobiphenyl, 3-Phenylaniline, m-Aminobiphenyl, m-Phenylaniline |
| InChI Formula | InChI=1S/C12H11N/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9H,13H2 |
| SMILES | Nc1cccc(c1)c2ccccc2 |
| IUPAC Name | 3-phenylaniline |
| Molecular Weight (g/mol) | 169.22 |
| Formula Weight | 169.0891 |