Learn More
3-Aminopyridine-2-carboxaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Aminopyridine-2-carboxaldehyde |
| CAS | 55234-58-7 |
| Molecular Formula | C6 H6 N2 O |
| Quantity | 50 mg |
| Synonym | 2-Pyridinecarboxaldehyde, 3-amino-, Picolinaldehyde, 3-amino- (6CI), 3-Amino-2-pyridinecarboxaldehyde, 3-Aminopicolinaldehyde, 3-Aminopyridine-2-carboxaldehyde |
| InChI Formula | InChI=1S/C6H6N2O/c7-5-2-1-3-8-6(5)4-9/h1-4H,7H2 |
| SMILES | Nc1cccnc1C=O |
| IUPAC Name | 3-aminopyridine-2-carbaldehyde |
| Molecular Weight (g/mol) | 122.12 |
| Formula Weight | 122.048 |