missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-58205
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6530-09-2
C7H14N2·2ClH
199.12
MFCD00137395
STZHBULOYDCZET-UHFFFAOYNA-N
This item is not returnable. View return policy
6530-09-2
C7H14N2·2ClH
199.12
MFCD00137395
STZHBULOYDCZET-UHFFFAOYNA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 321°C to 323°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Synonym 3-aminoquinuclidine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, quinuclidine, 3-amino-, dihydrochloride, 3aminoquinuclidine dihydrochloride, quinuclidin-3-amine dihydrochloride, 1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3-amino-1-azabicyclo 2.2.2 octane dihydrochloride, quinuclidine-3-ylamine, chloride, chloride
Solubility Information Solubility in water: soluble
SMILES C1CN2CCC1C(C2)N.Cl.Cl
IUPAC Name 1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
PubChem CID 197853
Formula Weight 199.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Aminoquinuclidine dihydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminoquinuclidine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'