missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminosulfonyl)thiophene-2-carboxylic Acid, TRC

Product Code. 30005670
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005670 50 mg 50mg
30138549 500 mg 500mg
30004069 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005670 Supplier Toronto Research Chemicals Supplier No. TRCA63302550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59337-97-2
Molecular Formula C5H5NO4S2
Molecular Weight (g/mol) 207.23
Synonym 3-(Aminosulfonyl)-2-thiophenecarboxylic acid (ACI), 3-Sulfamoyl-thiophene-2-carboxylic acid, 3-Sulfamoylthiophene-2-carboxylic acid, 2-Thiophenecarboxylic acid, 3-(aminosulfonyl)- (9CI, ACI), IN-L9223
IUPAC Name 3-sulfamoylthiophene-2-carboxylic acid
SMILES NS(=O)(=O)c1ccsc1C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H5NO4S2/c6-12(9,10)3-1-2-11-4(3)5(7)8/h1-2H,(H,7,8)(H2,6,9,10)
Formula Weight 206.966
Chemical Name or Material 3-(Aminosulfonyl)thiophene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.