Learn More
3-Benzyladenine, TRC
Product Code. 30134240
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134240 50 mg 50mg
30133434 5 mg 5mg
30008317 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134240 Supplier Toronto Research Chemicals Supplier No. TRCB13040850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7280-81-1
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.25
IUPAC Name 3-benzyl-7H-purin-6-imine
SMILES Nc1ncn(Cc2ccccc2)c3ncnc13
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H11N5/c13-11-10-12(15-7-14-10)17(8-16-11)6-9-4-2-1-3-5-9/h1-5,7-8,13H,6H2,(H,14,15)
Formula Weight 225.1014
Chemical Name or Material 3-Benzyladenine