Learn More
3-Benzyloxy-4-hydroxybenzaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Benzyloxy-4-hydroxybenzaldehyde |
| CAS | 50773-56-3 |
| Molecular Formula | C14H12O3 |
| Quantity | 1 g |
| Synonym | 4-Hydroxy-3-(phenylmethoxy)-benzaldehyde, 3-(Phenylmethoxy)-4-hydroxybenzaldehyde, 4-Hydroxy-3-(phenylmethoxy)benzaldehyde |
| InChI Formula | InChI=1S/C14H12O3/c15-9-12-6-7-13(16)14(8-12)17-10-11-4-2-1-3-5-11/h1-9,16H,10H2 |
| SMILES | Oc1ccc(C=O)cc1OCc2ccccc2 |
| IUPAC Name | 4-hydroxy-3-phenylmethoxybenzaldehyde |
| Molecular Weight (g/mol) | 228.24 |
| Formula Weight | 228.0786 |