missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzeneboronic acid, 98+%, Thermo Scientific™

Product Code. p-7047056
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11398436

Brand: Thermo Scientific Alfa Aesar L17474.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156682-54-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.05
MDL Number MFCD02093063
InChI Key WIJNYNBSPQMJGO-UHFFFAOYSA-N
Synonym 3-benzyloxyphenylboronic acid, 3-benzyloxybenzeneboronic acid, 3-benzyloxy phenyl boronic acid, 3-benzyloxy phenylboronic acid, m-benzyloxyphenylboronic acid, 3-benzyloxyphenyl boronic acid, 3-boronophenoxy methyl benzene, 3-benzyl-oxy phenyl boranediol, boronic acid, 3-phenylmethoxy phenyl
PubChem CID 2734313
IUPAC Name (3-phenylmethoxyphenyl)boronic acid
SMILES OB(O)C1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 122°C to 124°C
Quantity 1 g
Beilstein 6808244
Formula Weight 228.06
Percent Purity ≥98%
Chemical Name or Material 3-Benzyloxybenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.