missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylpyridine, 97%, ACROS Organics™

Brand:  Acros Organics 436860010

Additional Details : CAS : 38749-79-0 Weight : 0.05100kg


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 11976351

  • € 63.59 / 1g


Estimated Shipment: 18-08-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

38749-79-0
172.03
AIPWPTPHMIYYOX-UHFFFAOYSA-N
1201003
CC1=C(Br)C=CC=N1
C6H6BrN
MFCD00191224
3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
3-bromo-2-methylpyridine

Specifications

3-Bromo-2-methylpyridine
79°C
Glass bottle
65°C to 67°C (10.0 Torr)
1g
96.0
96% min. (GC)
MFCD00191224
AIPWPTPHMIYYOX-UHFFFAOYSA-N
3-bromo-2-methylpyridine
1201003
Liquid
1.4950g/mL
Authentic
1.495
Brown to Colorless
38749-79-0
100.0
C6H6BrN
3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
CC1=C(Br)C=CC=N1
172.03
172.02
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Bromo-2-methylpyridine, 97%

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates