missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylaniline, 97%, Thermo Scientific Chemicals

Catalog Number p-7027699
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7745-91-7
C7H8BrN
186.05
MFCD00134176
GRXMMIBZRMKADT-UHFFFAOYSA-N
3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline
82187
3-bromo-4-methylaniline
CC1=CC=C(N)C=C1Br
This item is not returnable. View return policy
7745-91-7
C7H8BrN
186.05
MFCD00134176
GRXMMIBZRMKADT-UHFFFAOYSA-N
3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline
82187
3-bromo-4-methylaniline
CC1=CC=C(N)C=C1Br

3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger?s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger′s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 25°C to 28°C
Boiling Point 254°C to 257°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.6115
Quantity 5 g
UN Number UN2811
Beilstein 1562057
Solubility Information Insoluble in water.
Formula Weight 186.06
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'