Learn More
3-Bromo-4-nitropyridine N-oxide, 98+%, Thermo Scientific Chemicals
Product Code. p-7047620
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318896 1 g 1g
11328896 5 g 5g
11338896 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11318896 Supplier Thermo Scientific Alfa Aesar Supplier No. L19524.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Sparingly Soluble in water (0.92 g/L at 25°C).
Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1678-49-5
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
MDL Number MFCD00085925
InChI Key YZQMKADIENBVLH-UHFFFAOYSA-N
Synonym 3-bromo-4-nitropyridine n-oxide, 3-bromo-4-nitropyridine 1-oxide, 3-bromo-4-nitropyridin-1-ium-1-olate, pyridine, 3-bromo-4-nitro-, 1-oxide, 3-bromo-4-nitropyridine-n-oxide, 3-bromo-4-nitro-1-oxido-pyridin-1-ium, 3-bromo-4-nitropyridin-1-ol, acmc-1bxbw, 3-bromo-4-nitropyridine oxide, 3-bromo-4-nitropyridin-n-oxide
PubChem CID 548521
IUPAC Name 3-bromo-4-nitro-1-oxidopyridin-1-ium
SMILES C1=C[N+](=CC(=C1[N+](=O)[O-])Br)[O-]
Specifications
Melting Point 152°C to 154°C
Quantity 1 g
Beilstein 150822
Solubility Information Sparingly Soluble in water (0.92g/L at 25°C).
Formula Weight 219
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-nitropyridine N-oxide