missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-methoxybenzyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64980.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439296

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

MFCD27975705

Specifications

3-Bromo-N-(4-methoxybenzyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27975705
Solid
1g
424.23
97%
Videos
Documents