missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldehyde ethylene acetal, 98+%

Product Code. 11305395
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305395 25 g 25g
11395385 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305395 Supplier Thermo Scientific Alfa Aesar Supplier No. L19611.14
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 17789-14-9 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17789-14-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00003209
InChI Key VYPYKCPWNPPBBX-UHFFFAOYSA-N
Synonym 2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
PubChem CID 87306
IUPAC Name 2-(3-bromophenyl)-1,3-dioxolane
SMILES C1COC(O1)C2=CC(=CC=C2)Br

Specifications

Density 1.514
Boiling Point 132°C to 133°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 25 g
Beilstein 1344030
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzaldehyde ethylene acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.