missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzamide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7055952
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
22726-00-7
C7H6BrNO
200.035
MFCD00017127
ODJFDWIECLJWSR-UHFFFAOYSA-N
m-bromobenzamide, benzamide, 3-bromo, 3-bromo-benzamide, benzamide, m-bromo, 3-carbamoyl bromobenzene, 3-bromanylbenzamide, pubchem3734, 3-bromophenylcarboxamide, acmc-209fyc, 3-bromobenzoic acid amide
89807
3-bromobenzamide
C1=CC(=CC(=C1)Br)C(=O)N
This item is not returnable. View return policy
22726-00-7
C7H6BrNO
200.035
MFCD00017127
ODJFDWIECLJWSR-UHFFFAOYSA-N
m-bromobenzamide, benzamide, 3-bromo, 3-bromo-benzamide, benzamide, m-bromo, 3-carbamoyl bromobenzene, 3-bromanylbenzamide, pubchem3734, 3-bromophenylcarboxamide, acmc-209fyc, 3-bromobenzoic acid amide
89807
3-bromobenzamide
C1=CC(=CC(=C1)Br)C(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 156°C to 159°C
Color White
Quantity 25 g
Beilstein 1860187
Formula Weight 200.04
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 3-Bromobenzamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'