Learn More
3-Butylphthalide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-n-Butylphthalide |
| CAS | 6066-49-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H14 O2 |
| Quantity | 25 g |
| Synonym | 3-Butyl-1(3H)-isobenzofuranone, 3-n-Butylphthalide |
| InChI Formula | InChI=1S/C12H14O2/c1-2-3-8-11-9-6-4-5-7-10(9)12(13)14-11/h4-7,11H,2-3,8H2,1H3 |
| SMILES | CCCCC1OC(=O)c2ccccc12 |
| IUPAC Name | 3-butyl-3H-2-benzofuran-1-one |
| Molecular Weight (g/mol) | 190.24 |
| Show More |