missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamoylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 444450050

Additional Details : CAS : 188665-74-9 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11986031

  • € 317.00 / 5g


In stock, ready to ship: 15
Add to basket

Description

Description

Specifications

Chemical Identifiers

188665-74-9
247.1
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide, 3-aminocarbonylphenylboronic acid pinacol ester, benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 2-dioxaborolan-2-yl-benzamide, bpkiphywhvowms-uhfffaoysa-n, 3-carbamoylphenylboronic acid pinacol ester
C13H18BNO3
BPKIPHYWHVOWMS-UHFFFAOYSA-N
2760587

Specifications

3-Carbamoylphenylboronic acid pinacol ester
5g
96% min. (GC)
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide, 3-aminocarbonylphenylboronic acid pinacol ester, benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 2-dioxaborolan-2-yl-benzamide, bpkiphywhvowms-uhfffaoysa-n, 3-carbamoylphenylboronic acid pinacol ester
247.1
247.1
182°C to 183°C
188665-74-9
C13H18BNO3
BPKIPHYWHVOWMS-UHFFFAOYSA-N
2760587
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Carbamoylphenylboronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates