missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-1,2-benzisoxazole, 97%, Thermo Scientific™

Product Code. 10376564
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10376564

Brand: Thermo Scientific Maybridge MO00450DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16263-52-8
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.565
MDL Number MFCD03407314
InChI Key INWUFXPCLZRSBH-UHFFFAOYSA-N
PubChem CID 2779780
IUPAC Name 3-chloro-1,2-benzoxazole
SMILES C1=CC=C2C(=C1)C(=NO2)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 3-chloro-1,2-benzisoxazole, 3-chlorobenzo d isoxazole, 1,2-benzisoxazole,3-chloro, 3-chloro-1,2-benzoisoxazole, 3-chloro-1,2-benzisooxazole, 3-chloranyl-1,2-benzoxazole, 3-chlorobenzisoxazole, pubchem8716, zlchem 1012, 1,2-benzisoxazole, 3-chloro
Formula Weight 153.57
Percent Purity 97%
Chemical Name or Material 3-chloro-1, 2-benzisoxazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.