Learn More
3-Chloro-2,4,5,6-tetrafluoropyridine, 98%, Thermo Scientific™
Product Code. 11433517
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433517 25 g 25g
11423517 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11433517 Supplier Thermo Scientific Alfa Aesar Supplier No. A17285.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1735-84-8
Molecular Formula C5ClF4N
Molecular Weight (g/mol) 185.51
MDL Number MFCD00010624
InChI Key UXUZMYHSICIOQT-UHFFFAOYSA-N
PubChem CID 137183
IUPAC Name 3-chloro-2,4,5,6-tetrafluoropyridine
SMILES FC1=NC(F)=C(Cl)C(F)=C1F
Specifications
Density 1.641
Boiling Point 118°C to 121°C
Flash Point >110°C (230°F)
Refractive Index 1.434
Quantity 25 g
UN Number UN1760
Beilstein 475873
Synonym pyridine, 3-chloro-2,4,5,6-tetrafluoro, 3-chlorotetrafluoropyridine, 2-chloro-2,4,5,6-tetrafluoropyridine, pubchem9226, 2,3,4,6-tetrafluoro-5-chloropyridine, 5-chloro-2,3,4,6-tetrafluoropyridine, 3-chloro-2,4,5,6-tetrafluoro-pyridine, pyridine,3-chloro-2,4,5,6-tetrafluoro, 3-chloro-2,4,5,6-tetrafluoropyridine, 3-chloranyl-2,4,5,6-tetrakis fluoranyl pyridine
Formula Weight 185.51
Percent Purity 98%
Chemical Name or Material 3-Chloro-2,4,5,6-tetrafluoropyridine