missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)phenylacetic acid, 97%

Product Code. 15497585
Change view
Click to view available options
Quantity:
250mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497585 250mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15497585 Supplier Alfa Aesar™ Supplier No. H26524.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 261763-13-7
Molecular Formula C9H5ClF4O2
MDL Number MFCD01631516
Quantity 250mg
Synonym 3-chloro-2-fluoro-5-trifluoromethyl phenylacetic acid, 2-3-chloro-2-fluoro-5-trifluoromethyl phenyl acetic acid, 3-chloro-2-fluoro-5-trifluoromethyl phenylaceticacid, 3-chloro-2-fluoro-5-trifluoromethyl phenyl acetic acid, 3-chloro-2-fluoro-5-trifluoromethyl-phenylacetic acid, pubchem7142, 3-chloro-2-fluoro-5-trifluoromethyl-phenyl-acetic acid
InChI Key ORXABTQTILJYQU-UHFFFAOYSA-N
SMILES OC(=O)CC1=C(F)C(Cl)=CC(=C1)C(F)(F)F
IUPAC Name 2-[3-chloro-2-fluoro-5-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol) 256.58
PubChem CID 2773772
Formula Weight 256.58
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)phenylacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.