missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-(3-Chloro-2-hydroxypropyl)trimethyl-d9-ammonium Chloride, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-2-hydroxypropyltrimethylammonium-D9 (N,N,N-trimethyl-D9) Chloride |
| Molecular Formula | C6 D9 H6 Cl N O . Cl |
| Quantity | 0.05 g |
| Synonym | (±)-(3-Chloro-2-hydroxypropyl)trimethyl-d9-ammonium Chloride, 3-Chloro-2-hydroxypropyltrimethylammonium-d9 chloride, 3-Chloro-2-hydroxypropyltri(methyl-d3)ammonium-d9 chloride, 3-Chloro-2-hydroxy-N,N,N-tri(methyl-d3)-1-propanaminium chloride, 3-Chloro-2-hydroxypropyltrimethylammonium-D9 (N,N,N-trimethyl-D9) Chloride |
| InChI Formula | InChI=1 S/C6H15ClNO.ClH/c1-8(2,3)5-6(9)4-7;/h6,9 H,4-5H2,1-3H3;1 H/q+1;/p-1/i1D3,2D3,3D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](CC(O)CCl)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | (3-chloro-2-hydroxypropyl)-tris(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 197.1509 |
| Formula Weight | 196.1096 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction