Learn More
3-Chloro-4-ethylphenol, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-4-ethylphenol |
| CAS | 1243290-06-3 |
| Molecular Formula | C8H9OCl |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C8H9ClO/c1-2-6-3-4-7(10)5-8(6)9/h3-5,10H,2H2,1H3 |
| SMILES | CCc1ccc(O)cc1Cl |
| IUPAC Name | 3-chloro-4-ethylphenol |
| Molecular Weight (g/mol) | 156.61 |
| Formula Weight | 156.0342 |