missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-nitrobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. p-7028710
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15493387 250 mg 250mg
11342099 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15493387

Brand: Thermo Scientific Alfa Aesar H26115.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-4-nitrobenzenesulfonyl chloride is the reactant in the synthesis of aminobenzenesulfonyl fluorides. 3-Chloro-4-nitrobenzenesulfonyl chloride is also used as intermediates in organic synthesis.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64835-30-9
Molecular Formula C6H3Cl2NO4S
Molecular Weight (g/mol) 256.05
MDL Number MFCD04973767
InChI Key BYKPVHSGMXQEEY-UHFFFAOYSA-N
Synonym 3-chloro-4-nitrobenzene-1-sulfonyl chloride, 3-chloro-4-nitrobenzenesulphonyl chloride, acmc-1bffq, 3-chloro-4-nitrobenzenesulfonylchloride, 3-chloro-4-nitrophenylsulfonyl chloride, 3-chloro-4-nitro-benzenesulfonyl chloride, 3-chloro-4-nitrobenzene sulfonyl chloride, benzenesulfonylchloride, 3-chloro-4-nitro, 3-chloranyl-4-nitro-benzenesulfonyl chloride
PubChem CID 44630751
SMILES [O-][N+](=O)C1=C(Cl)C=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 36°C to 39°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
IUPAC Name 3-chloro-4-nitrobenzenesulfonyl chloride
Formula Weight 256.07
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-nitrobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.