missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 11475667
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475667 2 g 2g
11485667 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475667 Supplier Thermo Scientific Alfa Aesar Supplier No. B23630.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-4-(trifluoromethoxy)aniline is used as a primary and secondary intermediate in chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64628-73-5
Molecular Formula C7H5ClF3NO
Molecular Weight (g/mol) 211.568
MDL Number MFCD00190126
InChI Key ZPKUUNGPBSRPRM-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethoxy aniline, benzenamine, 3-chloro-4-trifluoromethoxy, 3-chloro-4-trifluoromethoxy phenylamine, pubchem2949, acmc-1b4hb, ksc496e4r, 3-chloro4-trifluoromethoxyaniline, 3-chloro-4-trifluoromethoxy-aniline, 3-chloro-4-trifluoromethoxy-phenylamine
PubChem CID 2773843
IUPAC Name 3-chloro-4-(trifluoromethoxy)aniline
SMILES C1=CC(=C(C=C1N)Cl)OC(F)(F)F

Specifications

Melting Point 29°C to 30°C
Density 1.454
Quantity 2 g
Beilstein 6314199
Solubility Information Slightly soluble in water.
Formula Weight 211.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.