Learn More
3-Chloro-5-methoxybenzeneboronic acid, 94%, Thermo Scientific™
Product Code. p-7029920
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453687 250 mg 250mg
11357289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15453687 Supplier Thermo Scientific Alfa Aesar Supplier No. H27442.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as an active pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 915201-07-9
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.398
MDL Number MFCD03095046
InChI Key WAVZSRUGKKOQRB-UHFFFAOYSA-N
Synonym 3-chloro-5-methoxyphenyl boronic acid, 3-chloro-5-methoxybenzeneboronic acid, boronic acid, 3-chloro-5-methoxyphenyl, b-3-chloro-5-methoxyphenyl-boronic acid, boronic acid, b-3-chloro-5-methoxyphenyl, pubchem1777, acmc-209rc2, 3-chloro-5-methoxyphenylboronicacid, 3-chloro-5-methoxyphenylboronic acid
PubChem CID 17750234
IUPAC Name (3-chloro-5-methoxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)Cl)OC)(O)O
Specifications
Melting Point 185°C to 189°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 186.4
Percent Purity 94%
Chemical Name or Material 3-Chloro-5-methoxybenzeneboronic acid