Learn More
3-Chloro-6,11-dihydro-5,5-dioxo-11-hydroxy-6-methyldibenzo[c,f][1,2]thiazepine, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-6,11-dihydro-5,5-dioxo-11-hydroxy-6-methyldibenzo[c,f][1,2]thiazepine |
| CAS | 26723-60-4 |
| Molecular Formula | C14H12ClNO3S |
| Quantity | 25 mg |
| Synonym | 3-Chloro-6,11-dihydro-5,5-dioxo-11-hydroxy-6-methyldibenzo[c,f][1,2]thiazepine, 3-Chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-ol 5,5-dioxide, 3-Chloro-6-methyl-5,5-dioxo-11H-benzo[c][2,1]benzothiazepin-11-ol |
| InChI Formula | InChI=1S/C14H12ClNO3S/c1-16-12-5-3-2-4-10(12)14(17)11-7-6-9(15)8-13(11)20(16,18)19/h2-8,14,17H,1H3 |
| SMILES | CN1C2=C(C=CC=C2)C(O)C3=CC=C(Cl)C=C3S1(=O)=O |
| IUPAC Name | 3-chloro-11-hydroxy-6-methyl-6,11-dihydrodibenzo[c,f][1,2]thiazepine 5,5-dioxide |
| Molecular Weight (g/mol) | 309.77 |
| Formula Weight | 309.02 |