Learn More
3-Chloro-6-methyl-dibenzo[c,f][1,2]thiazepin-11(6H)-one 5,5-Dioxide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-6-methyl-dibenzo[c,f][1,2]thiazepin-11(6H)-one 5,5-dioxide |
| CAS | 26638-53-9 |
| Molecular Formula | C14 H10 Cl N O3 S |
| Quantity | 5 g |
| Synonym | Dibenzo[c,f][1,2]thiazepin-11(6H)-one, 3-chloro-6-methyl-, 5,5-dioxide, 3-Chloro-6,11-dihydro-6-methyl-5,5,11-trioxodibenzo[c,f][1,2]thiazepine, 3-Chloro-6-methyldibenzo[c,f][1,2]thiazepin-11(6H)-one S,S-Dioxide |
| InChI Formula | InChI=1S/C14H10ClNO3S/c1-16-12-5-3-2-4-10(12)14(17)11-7-6-9(15)8-13(11)20(16,18)19/h2-8H,1H3 |
| SMILES | CN1c2ccccc2C(=O)c3ccc(Cl)cc3S1(=O)=O |
| IUPAC Name | 3-chloro-6-methyl-5,5-dioxobenzo[c][1,2]benzothiazepin-11-one |
| Molecular Weight (g/mol) | 307.75 |
| Formula Weight | 307.007 |