Learn More
3-Chlorobenzotrifluoride, TRC
Code produit 30351347
Modifier l'affichage
Click to view available options
Quantity:
25 g
Conditionnement:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
30351347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30351347 Fournisseur Toronto Research Chemicals Code fournisseur TRCC42874525G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material 3-Chloro-alpha,alpha,alpha-trifluorotoluene
CAS 98-15-7
Molecular Formula C7 H4 Cl F3
Quantity 25 g
Synonym Fluoxetine Impurity 16
InChI Formula InChI=1S/C7H4ClF3/c8-6-3-1-2-5(4-6)7(9,10)11/h1-4H
SMILES FC(F)(F)c1cccc(Cl)c1
IUPAC Name 1-chloro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol) 180.55
Formula Weight 179.9954