Learn More
3-Chloropyridazine Hydrochloride, TRC
Product Code. 30348104
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348104 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30348104 Supplier Toronto Research Chemicals Supplier No. TRCB40523350MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloropyridazine Hydrochloride
CAS 856847-77-3
Molecular Formula C4H3N2Cl . HCl
Quantity 50 mg
InChI Formula InChI=1S/C4H3ClN2.ClH/c5-4-2-1-3-6-7-4;/h1-3H;1H
SMILES Cl.Clc1cccnn1
IUPAC Name 3-chloropyridazine;hydrochloride
Molecular Weight (g/mol) 150.99
Formula Weight 149.9752