Learn More
3-Cyano-4-fluorobenzaldehyde, TRC
Product Code. 30361031
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361031 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30361031 Supplier Toronto Research Chemicals Supplier No. TRCC9875151G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Cyano-4-fluorobenzaldehyde
CAS 218301-22-5
Molecular Formula C8H4FNO
Quantity 1 g
Synonym 2-Fluoro-5-formylbenzonitrile
InChI Formula InChI=1S/C8H4FNO/c9-8-2-1-6(5-11)3-7(8)4-10/h1-3,5H
SMILES Fc1ccc(C=O)cc1C#N
IUPAC Name 2-fluoro-5-formylbenzonitrile
Molecular Weight (g/mol) 149.12
Formula Weight 149.0277