missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals

Catalog Number p-7055646
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
21642-98-8
C7H6N2O2
150.14
MFCD00975442
MWGIDWPSRDMIQN-UHFFFAOYSA-N
4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile, 4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile, 3-cyano-2-hydroxy-4-methoxypyridine, 4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 2-hydroxy-4-methoxynicotinonitrile, 1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo, 3-cyano-4-methoxy-2 1h-pyridinone, 2-hydroxy-4-methoxy-nicotinonitrile, 3-cyano-4-methoxy-2-1h-pyridinone
2786702
COC1=C(C#N)C(=O)NC=C1
This item is not returnable. View return policy
21642-98-8
C7H6N2O2
150.14
MFCD00975442
MWGIDWPSRDMIQN-UHFFFAOYSA-N
4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile, 4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile, 3-cyano-2-hydroxy-4-methoxypyridine, 4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 2-hydroxy-4-methoxynicotinonitrile, 1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo, 3-cyano-4-methoxy-2 1h-pyridinone, 2-hydroxy-4-methoxy-nicotinonitrile, 3-cyano-4-methoxy-2-1h-pyridinone
2786702
COC1=C(C#N)C(=O)NC=C1

Used as a intermediate for gimeracil and pharmaceutical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a intermediate for gimeracil and pharmaceutical application.

Solubility
Soluble in Dimethyl sulfoxide.

Notes
Hydroscopic. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from water/moisture and oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 278°C
Color Yellow
Quantity 1 g
UN Number UN3439
Beilstein 5929290
Sensitivity Hygroscopic
Solubility Information Soluble in Dimethyl sulfoxide.
IUPAC Name 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formula Weight 150.14
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 3-Cyano-4-methoxy-2-pyridone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'