Learn More
3-Des[2-(Dimethylamino)ethyl] Zolmitriptan 3-Acetic Acid, TRC
Product Code. 30349817
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349817 5 mg 5mg
30355869 50 mg 50mg
30351675 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30349817 Supplier Toronto Research Chemicals Supplier No. TRCD2889655MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 251451-31-7
Molecular Formula C14 H14 N2 O4
Molecular Weight (g/mol) 274.27
IUPAC Name 2-[5-[[(4S)-2-oxo-1,3-oxazolidin-4-yl]methyl]-1H-indol-3-yl]acetic acid
SMILES OC(=O)Cc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C14H14N2O4/c17-13(18)5-9-6-15-12-2-1-8(4-11(9)12)3-10-7-20-14(19)16-10/h1-2,4,6,10,15H,3,5,7H2,(H,16,19)(H,17,18)/t10-/m0/s1
Formula Weight 274.0954
Chemical Name or Material 3-Des[2-(Dimethylamino)ethyl] Zolmitriptan 3-Acetic Acid